Assay Details
Target Gene Details
Entrez Gene ID: | 338811 |
Gene Name: | family with sequence similarity 19 member A2, C-C motif chemokine like |
Gene Aliases: |
TAFA-2, TAFA2 |
Location: |
Chr.12:61708248-62192839 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAM19A2 | NM_178539.4 | 5 | 2922 | NP_848634.1 |
| XM_006719370.2 | 6 | 2651 | XP_006719433.1 | |
| XM_011538272.1 | 6 | 2984 | XP_011536574.1 | |
| XM_011538274.1 | 6 | 2375 | XP_011536576.1 | |
| AK123580.1 | 6 | 1530 | ||
| AL834160.1 | 5 | 2274 | CAD38865.1 | |
| BC028403.1 | 5 | 1871 | AAH28403.1 | |
| BC040286.1 | 4 | 1314 | AAH40286.1 | |
| CR749367.1 | 9 | 2068 | CAH18220.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581660 | Chr.12:61685385 - 61733005 on Build GRCh38 | Gain |
|
| nsv1042044 | Chr.12:60862234 - 61803104 on Build GRCh38 | Gain |
|
| nsv520898 | Chr.12:61684989 - 61721384 on Build GRCh38 | Gain |
|
| nsv1039741 | Chr.12:61653069 - 62097271 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map