Assay Details
Target Gene Details
Entrez Gene ID: | 23155 |
Gene Name: | chloride channel CLIC like 1 |
Gene Aliases: |
MCLC |
Location: |
Chr.1:108929508-108963499 on Build GRCh38 |
Assay Gene Location: | Within Exon 12 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CLCC1 | NM_001048210.2 | 12 | 4215 | NP_001041675.1 |
| NM_001278202.1 | 9 | 3723 | NP_001265131.1 | |
| NM_001278203.1 | 7 | 3531 | NP_001265132.1 | |
| NM_015127.4 | 12 | 4075 | NP_055942.1 | |
| AB018304.2 | 11 | 4076 | BAA34481.2 | |
| AK090563.1 | 8 | 2279 |
Target Gene Details
Entrez Gene ID: | 29899 |
Gene Name: | G-protein signaling modulator 2 |
Gene Aliases: |
CMCS, DFNB82, LGN, PINS |
Location: |
Chr.1:108875350-108934335 on Build GRCh38 |
Assay Gene Location: | Within Exon 19 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPSM2 | NM_001321038.1 | 15 | 2507 | NP_001307967.1 |
| NM_001321039.1 | NP_001307968.1 | |||
| NM_013296.4 | 15 | 2739 | NP_037428.3 | |
| XM_006710589.2 | 14 | 3198 | XP_006710652.1 | |
| XM_011541301.2 | 16 | 3016 | XP_011539603.1 | |
| XM_011541302.2 | 16 | 3678 | XP_011539604.1 | |
| XM_017001097.1 | 16 | 3584 | XP_016856586.1 | |
| XM_017001098.1 | 16 | 3352 | XP_016856587.1 | |
| AB445462.1 | ||||
| AI224972.1 | 1 | 301 | ||
| AK295563.1 | 15 | 2323 | ||
| BC027732.2 | 15 | 2515 | AAH27732.1 | |
| KX085485.1 | 15 | 2739 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv15n21 | Chr.1:108873603 - 108941145 on Build GRCh38 | Gain |
|
| dgv251n100 | Chr.1:108930151 - 108955511 on Build GRCh38 | Loss |
|
| nsv831026 | Chr.1:108924711 - 109119614 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map