Assay Details
Target Gene Details
Entrez Gene ID: | 134728 |
Gene Name: | interleukin 1 receptor associated kinase 1 binding protein 1 |
Gene Aliases: |
AIP70, SIMPL |
Location: |
Chr.6:78867486-78979411 on Build GRCh38 |
Assay Gene Location: |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| IRAK1BP1 | AK309003.1 |
Target Gene Details
Entrez Gene ID: | 55023 |
Gene Name: | pleckstrin homology domain interacting protein |
Gene Aliases: |
BRWD2, DCAF14, WDR11, ndrp |
Location: |
Chr.6:78934419-79078294 on Build GRCh38 |
Assay Gene Location: | Within Exon 42 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PHIP | NM_017934.5 | 40 | 5371 | NP_060404.3 |
| XM_005248729.4 | 40 | 5368 | XP_005248786.1 | |
| XM_011535918.2 | 37 | 5027 | XP_011534220.1 | |
| XM_017010989.1 | 42 | 5513 | XP_016866478.1 | |
| XM_017010990.1 | 39 | 5111 | XP_016866479.1 | |
| AF310250.1 | 17 | 2378 | AAG45145.1 | |
| AJ303102.1 | 40 | 5158 | CAC83118.1 | |
| AK000712.1 | 13 | 2001 | BAA91336.1 | |
| AK075124.1 | 9 | 1458 | BAC11417.1 | |
| AL161957.1 | 17 | 5226 | CAH10776.1 | |
| BC008909.2 | 15 | 2099 | AAH08909.2 | |
| BC081569.1 | 18 | 2414 | AAH81569.1 | |
| BC137488.1 | 40 | 5251 | ||
| DQ924532.1 | 40 | 5145 |
More Information
Set Membership: |
|

Genomic Map