Assay Details
Target Gene Details
Entrez Gene ID: | 6455 |
Gene Name: | SH3 domain containing GRB2 like 1, endophilin A2 |
Gene Aliases: |
CNSA1, EEN, SH3D2B, SH3P8 |
Location: |
Chr.19:4360367-4400568 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 9 - Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SH3GL1 | NM_001199943.1 | NP_001186872.1 | ||
| NM_001199944.1 | NP_001186873.1 | |||
| NM_003025.3 | NP_003016.1 | |||
| XM_011528183.2 | XP_011526485.1 | |||
| XM_017027146.1 | XP_016882635.1 | |||
| AK097616.1 | ||||
| AK299166.1 | ||||
| AK315450.1 | ||||
| AY364240.1 | AAQ76799.1 | |||
| BC001270.1 | AAH01270.1 | |||
| BC098565.1 | AAH98565.1 | |||
| CR542087.1 | CAG46884.1 | |||
| U65999.1 | AAB86800.1 | |||
| X99656.1 | CAA67970.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv458328 | Chr.19:4320221 - 4386355 on Build GRCh38 | Gain |
|
| nsv833717 | Chr.19:4204757 - 4368519 on Build GRCh38 | Loss |
|
| nsv833719 | Chr.19:4325245 - 4378052 on Build GRCh38 | Loss |
|
| nsv833718 | Chr.19:4221577 - 4480248 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map