Assay Details
Target Gene Details
Entrez Gene ID: | 204801 |
Gene Name: | NLR family pyrin domain containing 11 |
Gene Aliases: |
CLR19.6, NALP11, NOD17, PAN10, PYPAF6, PYPAF7 |
Location: |
Chr.19:55785397-55836762 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NLRP11 | NM_001297743.1 | 2 | 678 | NP_001284672.1 |
| NM_145007.3 | 5 | 1528 | NP_659444.2 | |
| AK310394.1 | 5 | 1205 | ||
| AY095145.1 | 3 | 916 | AAM14632.1 | |
| AY154466.1 | 2 | 817 | AAO18162.1 | |
| AY168967.1 | 3 | 905 | AAO12532.1 | |
| AY168968.1 | 2 | 577 | AAO12533.1 | |
| AY226382.1 | 2 | 817 | AAO59428.1 | |
| BC034730.1 | 3 | 905 | AAH34730.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv833882 | Chr.19:55647267 - 55815198 on Build GRCh38 | Loss |
|
| esv2758773 | Chr.19:55724171 - 55814940 on Build GRCh38 | Gain |
|
| nsv580346 | Chr.19:55735154 - 55857948 on Build GRCh38 | Gain |
|
| dgv6582n54 | Chr.19:55807428 - 56027874 on Build GRCh38 | Gain |
|
| nsv1060269 | Chr.19:55708879 - 55810275 on Build GRCh38 | Gain |
|
| nsv1064105 | Chr.19:55808050 - 55939166 on Build GRCh38 | Gain |
|
| esv3893253 | Chr.19:55805430 - 55857497 on Build GRCh38 | Loss |
|
| nsv1057383 | Chr.19:55699665 - 55944662 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map