Assay Details
Target Gene Details
Entrez Gene ID: | 4008 |
Gene Name: | LIM domain 7 |
Gene Aliases: |
FBX20, FBXO20, LOMP |
Location: |
Chr.13:75620434-75859870 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO7 | NM_005358.5 | 1 | 261 | NP_005349.3 |
| AF330045.1 | 1 | 261 | AAL37480.1 |
Target Gene Details
Entrez Gene ID: | 101927155 |
Gene Name: | LMO7 antisense RNA 1 |
Gene Aliases: |
UCHL3-AS1 |
Location: |
Chr.13:75604700-75635994 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LMO7-AS1 | NR_120410.1 | |||
| NR_120411.1 | ||||
| NR_120412.1 | ||||
| AI816891.1 | ||||
| AW188034.1 | ||||
| BF593247.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2751152 | Chr.13:75570063 - 75714282 on Build GRCh38 | Loss |
|
| nsv521444 | Chr.13:74139302 - 75679218 on Build GRCh38 | Gain |
|
| esv3632750 | Chr.13:75615888 - 75621625 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map