Assay Details
Target Gene Details
Entrez Gene ID: | 1810 |
Gene Name: | down-regulator of transcription 1 |
Gene Aliases: |
NC2, NC2-BETA, NC2B |
Location: |
Chr.1:93345921-93362592 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DR1 | NM_001938.2 | 3 | 1195 | NP_001929.1 |
| AK057541.1 | 4 | 1057 | ||
| AK223442.1 | 3 | 1209 | BAD97162.1 | |
| AK311288.1 | 3 | 353 | ||
| AL137673.1 | 2 | 887 | ||
| AL833729.1 | 3 | 1178 | CAH56250.1 | |
| BC002809.2 | 3 | 1172 | AAH02809.1 | |
| BC035507.2 | 3 | 1195 | AAH35507.1 | |
| BC068553.1 | 3 | 1194 | AAH68553.1 | |
| BT006972.1 | 3 | 470 | AAP35618.1 | |
| M97388.1 | 4 | 1017 | AAA58442.1 |
More Information
Set Membership: |
|

Genomic Map