Assay Details
Target Gene Details
Entrez Gene ID: | 429 |
Gene Name: | achaete-scute family bHLH transcription factor 1 |
Gene Aliases: |
ASH1, HASH1, MASH1, bHLHa46 |
Location: |
Chr.12:102957674-102960516 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCL1 | NM_004316.3 | 2 | 2197 | NP_004307.2 |
| AW071836.1 | 1 | 294 | ||
| BC004425.1 | 2 | 2185 | AAH04425.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3581838 | Chr.12:102959348 - 102960527 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map