Assay Details
Target Gene Details
Entrez Gene ID: | 23462 |
Gene Name: | hes related family bHLH transcription factor with YRPW motif 1 |
Gene Aliases: |
BHLHb31, CHF2, HERP2, HESR1, HRT-1, OAF1, hHRT1 |
Location: |
Chr.8:79764010-79767863 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| HEY1 | NM_001040708.1 | 5 | 1118 | NP_001035798.1 |
| NM_001282851.1 | 2 | 848 | NP_001269780.1 | |
| NM_012258.3 | 5 | 1106 | NP_036390.3 | |
| AF151522.1 | 5 | 930 | AAD38967.1 | |
| AF176422.1 | 5 | 1007 | AAD51749.1 | |
| AF232239.1 | 5 | 1012 | AAF37297.1 | |
| AF254637.1 | 5 | 1023 | AAL06278.1 | |
| AF311883.1 | AAG31156.1 | |||
| AJ272214.1 | 5 | 991 | CAB75715.1 | |
| AK074641.1 | 5 | 1009 | ||
| AK092437.1 | 5 | 1025 | BAC03890.1 | |
| AK226041.1 | 5 | 1013 | ||
| AK293646.1 | 2 | 848 | ||
| AK307622.1 | 2 | 702 | ||
| AK313271.1 | ||||
| BC001873.1 | 5 | 1003 | AAH01873.1 | |
| BT020065.1 | AAV38868.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv7245n100 | Chr.8:79748262 - 79816274 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map