Assay Details
Target Gene Details
Entrez Gene ID: | 7374 |
Gene Name: | uracil DNA glycosylase |
Gene Aliases: |
DGU, HIGM4, HIGM5, UDG, UNG1, UNG15, UNG2 |
Location: |
Chr.12:109097594-109110993 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 3 - Intron 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| UNG | NM_003362.3 | NP_003353.1 | ||
| NM_080911.2 | NP_550433.1 | |||
| AK291341.1 | ||||
| AK299416.1 | ||||
| AK313552.1 | ||||
| BC015205.1 | AAH15205.1 | |||
| BC050634.2 | AAH50634.1 | |||
| BG326541.1 | ||||
| BQ950839.1 | ||||
| CR541673.1 | CAG46474.1 | |||
| CR749342.1 | CAH18195.1 | |||
| X15653.1 | CAA33679.1 | |||
| Y09008.1 | CAA70211.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv526106 | Chr.12:109098754 - 109104587 on Build GRCh38 | Loss |
|
| dgv1553n100 | Chr.12:109064624 - 109159352 on Build GRCh38 | Gain |
|
| esv2422361 | Chr.12:108792678 - 109118360 on Build GRCh38 | Duplication |
|
More Information
Set Membership: |
|

Genomic Map