Assay Details
Target Gene Details
Entrez Gene ID: | 3739 |
Gene Name: | potassium voltage-gated channel subfamily A member 4 |
Gene Aliases: |
HBK4, HK1, HPCN2, HUKII, KCNA4L, KCNA8, KV1.4, PCN2 |
Location: |
Chr.11:30009741-30017030 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KCNA4 | NM_002233.3 | 2 | 2500 | NP_002224.1 |
| AK313662.1 | 1 | 1779 | ||
| L02751.1 | 1 | 1259 | AAA36140.1 | |
| M55514.1 | 2 | 2411 | AAA60034.1 | |
| M60450.1 | 1 | 1556 | AAA61275.1 |
More Information
Set Membership: |
|

Genomic Map