Assay Details
Target Gene Details
Entrez Gene ID: | 57826 |
Gene Name: | RAP2C, member of RAS oncogene family |
Gene Aliases: |
- |
Location: |
Chr.X:132203024-132219480 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| RAP2C | NM_001271186.1 | 6 | 1508 | NP_001258115.1 |
| NM_001271187.1 | 6 | 843 | NP_001258116.1 | |
| NM_021183.4 | 4 | 1436 | NP_067006.3 | |
| NR_073150.1 | 7 | 884 | ||
| XM_006724775.2 | 5 | 1166 | XP_006724838.1 | |
| XM_011531376.1 | 6 | 1576 | XP_011529678.1 | |
| AK124801.1 | 3 | 746 | ||
| AK310997.1 | 7 | 851 | ||
| AL049685.2 | 3 | 668 | CAB41256.1 | |
| AL832461.1 | 3 | 987 | ||
| AY298955.1 | 4 | 1436 | AAP55684.1 | |
| BC003403.1 | 3 | 722 | AAH03403.1 | |
| BI767215.1 | 5 | 730 | ||
| BX399437.2 | 6 | 880 | ||
| BX647223.1 | 4 | 725 | ||
| CN308919.1 | 2 | 325 |
More Information
Set Membership: |
|

Genomic Map