Assay Details
Target Gene Details
Entrez Gene ID: | 23026 |
Gene Name: | myosin XVI |
Gene Aliases: |
MYAP3, MYR8, Myo16b, NYAP3, PPP1R107 |
Location: |
Chr.13:108495729-109208007 on Build GRCh38 |
Assay Gene Location: | Within Exon 35 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO16 | NM_001198950.1 | 33 | 5835 | NP_001185879.1 |
| NM_015011.1 | 33 | 5273 | NP_055826.1 | |
| XM_011521062.1 | 33 | 5381 | XP_011519364.1 | |
| AB020672.2 | 33 | 5273 | ||
| AB290159.1 | 33 | 5836 | ||
| BC146791.1 | 33 | 5273 |
Target Gene Details
Entrez Gene ID: | 100885782 |
Gene Name: | MYO16 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.13:109163905-109167303 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MYO16-AS1 | NR_047700.1 | |||
| BG181984.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv832718 | Chr.13:109076335 - 109249674 on Build GRCh38 | Gain |
|
| nsv563071 | Chr.13:108767490 - 109327122 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map