Assay Details
Target Gene Details
Entrez Gene ID: | 429 |
Gene Name: | achaete-scute family bHLH transcription factor 1 |
Gene Aliases: |
ASH1, HASH1, MASH1, bHLHa46 |
Location: |
Chr.12:102957674-102960516 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ASCL1 | NM_004316.3 | 1 | 1238 | NP_004307.2 |
| AK290539.1 | 1 | 1238 | ||
| AK314114.1 | 1 | 851 | ||
| BC002341.2 | 1 | 1219 | AAH02341.1 | |
| BC003134.1 | 1 | 1219 | AAH03134.1 | |
| BC004425.1 | 1 | 1226 | AAH04425.1 | |
| BC031299.1 | 1 | 1204 | AAH31299.1 | |
| CF454566.1 | ||||
| L08424.1 | 1 | 1099 | AAA58376.1 |
More Information
Set Membership: |
|

Genomic Map