Assay Details
Target Gene Details
Entrez Gene ID: | 55090 |
Gene Name: | mediator complex subunit 9 |
Gene Aliases: |
MED25 |
Location: |
Chr.17:17476986-17493220 on Build GRCh38 |
Assay Gene Location: | Within Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MED9 | NM_018019.2 | 2 | 1229 | NP_060489.1 |
| AK001055.1 | 2 | 1229 | BAA91483.1 | |
| BC104988.1 | 2 | 1207 | AAI04989.1 | |
| BC112029.1 | 2 | 1207 | AAI12030.1 |
More Information
Additional Information:
For this assay, SNP(s) [rs78649356] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map