Assay Details
Target Gene Details
Entrez Gene ID: | 84449 |
Gene Name: | zinc finger protein 333 |
Gene Aliases: |
- |
Location: |
Chr.19:14689787-14733746 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 10 - Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ZNF333 | NM_001300912.1 | NP_001287841.1 | ||
| NM_032433.2 | NP_115809.1 | |||
| XM_005260098.3 | XP_005260155.1 | |||
| XM_006722924.2 | XP_006722987.1 | |||
| XM_011528362.2 | XP_011526664.1 | |||
| XM_011528363.2 | XP_011526665.1 | |||
| XM_011528365.2 | XP_011526667.1 | |||
| XM_011528366.2 | XP_011526668.1 | |||
| XM_011528367.2 | XP_011526669.1 | |||
| XM_011528368.2 | XP_011526670.1 | |||
| XM_011528369.2 | XP_011526671.1 | |||
| XM_017027367.1 | XP_016882856.1 | |||
| XM_017027368.1 | XP_016882857.1 | |||
| XM_017027369.1 | XP_016882858.1 | |||
| XM_017027370.1 | XP_016882859.1 | |||
| XM_017027371.1 | XP_016882860.1 | |||
| AF372702.1 | AAM15528.1 | |||
| AK094028.1 | ||||
| AK291471.1 | ||||
| AK307147.1 | ||||
| AK309208.1 | ||||
| AL832544.1 | 1 | 59 | ||
| BC048107.1 | AAH48107.1 | |||
| BC064571.1 | 7 | 1162 | AAH64571.1 | |
| BX647475.1 | CAI56712.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv578680 | Chr.19:14695350 - 14735523 on Build GRCh38 | Loss |
|
| nsv1060306 | Chr.19:14697771 - 14721224 on Build GRCh38 | Loss |
|
| esv3893167 | Chr.19:14610447 - 14950404 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map