Assay Details
Target Gene Details
Entrez Gene ID: | 2880 |
Gene Name: | glutathione peroxidase 5 |
Gene Aliases: |
HEL-S-75p |
Location: |
Chr.6:28526012-28534951 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GPX5 | NM_001509.2 | NP_001500.1 | ||
| NM_003996.3 | NP_003987.2 | |||
| AJ005277.1 | CAA06463.1 | |||
| AK093099.1 | ||||
| BC128159.1 | ||||
| BC128160.1 | ||||
| FJ460514.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1074388 | Chr.6:28510222 - 29115423 on Build GRCh38 | Deletion |
|
| dgv3296n106 | Chr.6:28510223 - 28623423 on Build GRCh38 | Deletion |
|
| nsv601211 | Chr.6:28515705 - 28553539 on Build GRCh38 | Loss |
|
| esv3576110 | Chr.6:28449711 - 28681348 on Build GRCh38 | Gain |
|
| nsv601210 | Chr.6:28449445 - 28606638 on Build GRCh38 | Gain |
|
| nsv830621 | Chr.6:28506174 - 28707932 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map