Assay Details
Target Gene Details
Entrez Gene ID: | 57380 |
Gene Name: | MRS2, magnesium transporter |
Gene Aliases: |
HPT, MRS2L |
Location: |
Chr.6:24402908-24426194 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 7 - Intron 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MRS2 | NM_001286264.1 | NP_001273193.1 | ||
| NM_001286265.1 | NP_001273194.1 | |||
| NM_001286266.1 | NP_001273195.1 | |||
| NM_020662.3 | NP_065713.1 | |||
| NR_104423.1 | ||||
| XM_005249242.1 | XP_005249299.1 | |||
| AF288288.1 | AAG01170.1 | |||
| AF293076.1 | AAK38615.1 | |||
| AK054587.1 | ||||
| AK291058.1 | ||||
| AK297418.1 | ||||
| AK298849.1 | ||||
| AK299825.1 | ||||
| BC001028.2 | AAH01028.2 | |||
| BC069009.1 | AAH69009.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1026638 | Chr.6:24347744 - 24475072 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map