Assay Details
Target Gene Details
Entrez Gene ID: | 100132273 |
Gene Name: | NDUFA6 antisense RNA 1 (head to head) |
Gene Aliases: |
- |
Location: |
Chr.22:42090933-42125349 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NDUFA6-AS1 | NR_034118.2 | |||
| AK123630.1 | ||||
| DA317430.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv508737 | Chr.22:42052007 - 42134336 on Build GRCh38 | Insertion |
|
| nsv834210 | Chr.22:42057329 - 42209100 on Build GRCh38 | Loss |
|

Genomic Map