Assay Details
Target Gene Details
Entrez Gene ID: | 132612 |
Gene Name: | adenosine deaminase domain containing 1 |
Gene Aliases: |
Tenr |
Location: |
Chr.4:122378966-122429815 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADAD1 | NM_001159285.1 | 4 | 617 | NP_001152757.1 |
| NM_001159295.1 | 4 | 681 | NP_001152767.1 | |
| NM_139243.3 | 5 | 695 | NP_640336.1 | |
| XM_005262741.2 | 4 | 716 | XP_005262798.1 | |
| XM_005262743.2 | 4 | 616 | XP_005262800.1 | |
| XM_005262744.2 | 4 | 716 | XP_005262801.1 | |
| XM_005262745.2 | 4 | 716 | XP_005262802.1 | |
| XM_011531603.1 | 4 | 614 | XP_011529905.1 | |
| AK057303.1 | 5 | 649 | BAB71416.1 | |
| AK093297.1 | 4 | 605 | BAC04125.1 | |
| BC040229.1 | 4 | 563 | AAH40229.1 | |
| DB058857.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3894021 | Chr.4:122306958 - 122473886 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map