Assay Details
Target Gene Details
Entrez Gene ID: | 374454 |
Gene Name: | keratin 77 |
Gene Aliases: |
K1B, KRT1B |
Location: |
Chr.12:52689612-52703524 on Build GRCh38 |
Assay Gene Location: | Within Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KRT77 | NM_175078.2 | NP_778253.2 | ||
| XM_011538288.2 | XP_011536590.1 | |||
| XM_011538289.2 | 5 | 1119 | XP_011536591.1 | |
| AJ564104.1 | CAD91892.1 | |||
| BC118598.1 | ||||
| BC122558.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv428281 | Chr.12:52570654 - 52761443 on Build GRCh38 | Gain |
|
| nsv832415 | Chr.12:52567563 - 52757300 on Build GRCh38 | Loss |
|
| esv3629524 | Chr.12:52688070 - 52835680 on Build GRCh38 | Gain |
|
| dgv2637n54 | Chr.12:52692676 - 52696406 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map