Assay Details
Target Gene Details
Entrez Gene ID: | 286676 |
Gene Name: | immunoglobulin like domain containing receptor 1 |
Gene Aliases: |
DFNB42, ILDR1alpha, ILDR1alpha', ILDR1beta |
Location: |
Chr.3:121987323-122060554 on Build GRCh38 |
Assay Gene Location: | Within Exon 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ILDR1 | NM_001199799.1 | 7 | 1058 | NP_001186728.1 |
| NM_001199800.1 | 5 | 791 | NP_001186729.1 | |
| NM_175924.3 | 6 | 926 | NP_787120.1 | |
| XM_005247389.4 | 5 | 1338 | XP_005247446.1 | |
| XM_011512738.2 | 7 | 1038 | XP_011511040.1 | |
| XM_011512739.2 | 6 | 800 | XP_011511041.1 | |
| AK129974.1 | 6 | 704 | BAC85264.1 | |
| AY134857.1 | 7 | 957 | AAN10256.1 | |
| AY672837.1 | 7 | 900 | AAT77412.1 | |
| AY672838.1 | 5 | 805 | AAT77413.1 | |
| AY672839.1 | 5 | 633 | AAT77414.1 | |
| BC044240.1 | 6 | 829 | AAH44240.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv33139 | Chr.3:121781553 - 122372984 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map