Assay Details
Target Gene Details
Entrez Gene ID: | 83932 |
Gene Name: | SprT-like N-terminal domain |
Gene Aliases: |
C1orf124, DVC1, PRO4323, spartan |
Location: |
Chr.1:231337936-231355029 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SPRTN | NM_001010984.3 | 4 | 1738 | NP_001010984.1 |
| NM_001261462.2 | 3 | 1609 | NP_001248391.1 | |
| NM_032018.6 | NP_114407.3 | |||
| XM_006711818.2 | XP_006711881.1 | |||
| XM_011544289.2 | XP_011542591.1 | |||
| AK027317.1 | 3 | 1280 | BAB55037.1 | |
| AK027613.1 | BAB55232.1 | |||
| AL512744.1 | CAC21670.1 | |||
| AY358611.1 | 4 | 2571 | AAQ88974.1 | |
| BC068478.1 | AAH68478.1 |
More Information
Set Membership: |
|

Genomic Map