Assay Details
Target Gene Details
Entrez Gene ID: | 9057 |
Gene Name: | solute carrier family 7 member 6 |
Gene Aliases: |
LAT-2, LAT3, y+LAT-2 |
Location: |
Chr.16:68263830-68301823 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A6 | NM_001076785.2 | 12 | 5901 | NP_001070253.1 |
| NM_003983.5 | 11 | 5814 | NP_003974.3 | |
| XM_011523433.1 | 11 | 5916 | XP_011521735.1 | |
| XM_011523434.1 | 11 | 5981 | XP_011521736.1 | |
| XM_011523438.1 | 10 | 6371 | XP_011521740.1 | |
| XM_011523439.1 | 12 | 6613 | XP_011521741.1 | |
| XM_017023851.1 | 10 | 5941 | XP_016879340.1 | |
| CR749291.1 | 11 | 5800 | CAH18146.1 | |
| CR749475.1 | 11 | 4624 | CAH18305.1 | |
| D87432.1 | 11 | 5849 |
Target Gene Details
Entrez Gene ID: | 84138 |
Gene Name: | solute carrier family 7 member 6 opposite strand |
Gene Aliases: |
- |
Location: |
Chr.16:68300615-68310965 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC7A6OS | NM_032178.2 | 5 | 876 | NP_115554.2 |
| AK023353.1 | 5 | 856 | BAB14538.1 | |
| AK091674.1 | 4 | 978 | ||
| AK225712.1 | 5 | 857 | ||
| AY078070.1 | 5 | 864 | AAL79958.1 | |
| BC013778.1 | 5 | 842 | AAH13778.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv510691 | Chr.16:68266527 - 68325431 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map