Assay Details
Target Gene Details
Entrez Gene ID: | 84172 |
Gene Name: | RNA polymerase I subunit B |
Gene Aliases: |
RPA135, RPA2, Rpo1-2 |
Location: |
Chr.2:112541915-112577153 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 2 - Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POLR1B | NM_001137604.2 | NP_001131076.1 | ||
| NM_001282772.1 | NP_001269701.1 | |||
| NM_001282774.1 | NP_001269703.1 | |||
| NM_001282776.1 | NP_001269705.1 | |||
| NM_001282777.1 | NP_001269706.1 | |||
| NM_001282779.1 | NP_001269708.1 | |||
| NM_019014.5 | NP_061887.2 | |||
| AK001678.1 | BAA91831.1 | |||
| AK022533.1 | BAB14082.1 | |||
| AK128044.1 | BAC87247.1 | |||
| AK294027.1 | ||||
| AK301178.1 | ||||
| AK302814.1 | ||||
| BC110833.1 | AAI10834.1 | |||
| BX647360.1 | ||||
| BX647683.1 |
More Information
Set Membership: |
|

Genomic Map