Assay Details
Target Gene Details
Entrez Gene ID: | 101927380 |
Gene Name: | C9orf41 antisense RNA 1 |
Gene Aliases: |
- |
Location: |
Chr.9:74952965-74997382 on Build GRCh38 |
Assay Gene Location: | Within Intron 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| C9orf41-AS1 | NR_121183.1 | |||
| BI552900.1 |
Target Gene Details
Entrez Gene ID: | 138199 |
Gene Name: | carnosine N-methyltransferase 1 |
Gene Aliases: |
C9orf41, UPF0586 |
Location: |
Chr.9:74980794-75028456 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CARNMT1 | NM_001320497.1 | 8 | 1435 | NP_001307426.1 |
| NM_152420.2 | 8 | 1755 | NP_689633.1 | |
| NR_135282.1 | 7 | 1614 | ||
| XM_005251701.3 | 8 | 1430 | XP_005251758.2 | |
| XM_017014269.1 | 8 | 1742 | XP_016869758.1 | |
| XM_017014270.1 | 9 | 1688 | XP_016869759.1 | |
| XM_017014271.1 | 9 | 2180 | XP_016869760.1 | |
| BC034033.1 | 8 | 1719 | AAH34033.1 | |
| BX538061.1 | 7 | 1272 | CAD97992.1 | |
| BX647386.1 | 8 | 1524 |
More Information
Set Membership: |
|

Genomic Map