Assay Details
Target Gene Details
Entrez Gene ID: | 23648 |
Gene Name: | single stranded DNA binding protein 3 |
Gene Aliases: |
CSDP, SSDP, SSDP1 |
Location: |
Chr.1:54225431-54413479 on Build GRCh38 |
Assay Gene Location: | Within Exon 21 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SSBP3 | NM_001009955.3 | 17 | 1773 | NP_001009955.1 |
| NM_018070.4 | 17 | 1794 | NP_060540.2 | |
| NM_145716.3 | 18 | 1854 | NP_663768.1 | |
| XM_006710545.3 | 16 | 1353 | XP_006710608.1 | |
| XM_017000897.1 | 18 | 1523 | XP_016856386.1 | |
| XM_017000898.1 | 15 | 1750 | XP_016856387.1 | |
| XM_017000899.1 | 17 | 1454 | XP_016856388.1 | |
| AF086085.1 | 4 | 492 | ||
| AI565975.1 | 1 | 310 | ||
| AK001217.1 | 14 | 1202 | BAA91561.1 | |
| AK093975.1 | 14 | 1514 | ||
| AK289474.1 | 17 | 1395 | ||
| BC004335.1 | 11 | 951 | AAH04335.2 | |
| BC066365.1 | 18 | 1598 | AAH66365.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1112532 | Chr.1:54224527 - 54227227 on Build GRCh38 | Deletion |
|
| nsv1078412 | Chr.1:54226526 - 54227227 on Build GRCh38 | Deletion |
|
More Information
Set Membership: |
|

Genomic Map