Assay Details
Target Gene Details
Entrez Gene ID: | 4744 |
Gene Name: | neurofilament heavy polypeptide |
Gene Aliases: |
CMT2CC, NFH |
Location: |
Chr.22:29480192-29491290 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 1 - Exon 2 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NEFH | NM_021076.3 | NP_066554.2 | ||
| XM_011530200.2 | XP_011528502.1 | |||
| AB020652.2 | ||||
| AF203032.1 | AAF13722.1 | |||
| AK302660.1 | ||||
| BC073969.1 | AAH73969.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3647536 | Chr.22:29477608 - 29543952 on Build GRCh38 | Gain |
|
| nsv1062039 | Chr.22:29458872 - 29493212 on Build GRCh38 | Gain |
|
| nsv438344 | Chr.22:29460090 - 29490054 on Build GRCh38 | Loss |
|
| esv3893473 | Chr.22:29474962 - 29528836 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map