Assay Details
Target Gene Details
Entrez Gene ID: | 51280 |
Gene Name: | golgi membrane protein 1 |
Gene Aliases: |
C9orf155, GOLPH2, GP73, HEL46, PSEC0257, bA379P1.3 |
Location: |
Chr.9:86026141-86100201 on Build GRCh38 |
Assay Gene Location: | Within Exon 11 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| GOLM1 | NM_016548.3 | 10 | 3002 | NP_057632.2 |
| NM_177937.2 | 10 | 2994 | NP_808800.1 | |
| AF236056.1 | 10 | 2943 | AAF44663.1 | |
| AI929234.1 | 1 | 89 | ||
| AJ420432.1 | 1 | 1225 | ||
| AK027717.1 | 10 | 2965 | ||
| AK075542.1 | 10 | 2936 | BAC11685.1 | |
| AK075557.1 | 10 | 2906 | ||
| AK092943.1 | 11 | 2882 | ||
| BC001740.1 | 10 | 2917 | AAH01740.1 | |
| CA446701.1 | 1 | 99 | ||
| EU794616.1 | 10 | 2961 |
Target Gene Details
Entrez Gene ID: | 60560 |
Gene Name: | N(alpha)-acetyltransferase 35, NatC auxiliary subunit |
Gene Aliases: |
EGAP, MAK10, MAK10P, bA379P1.1 |
Location: |
Chr.9:85941142-86026355 on Build GRCh38 |
Assay Gene Location: | Within Exon 25 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NAA35 | XM_011518903.2 | 20 | 6385 | XP_011517205.1 |
| XM_011518904.2 | 17 | 6071 | XP_011517206.1 | |
| XM_017015012.1 | 24 | 6641 | XP_016870501.1 |
More Information
Set Membership: |
|

Genomic Map