Assay Details
Target Gene Details
Entrez Gene ID: | 8576 |
Gene Name: | serine/threonine kinase 16 |
Gene Aliases: |
KRCT, MPSK, PKL12, TSF1 |
Location: |
Chr.2:219245469-219250337 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| STK16 | NM_001008910.2 | NP_001008910.1 | ||
| NR_026909.1 | 1 | 205 | ||
| XM_011512067.2 | XP_011510369.1 | |||
| XM_017005160.1 | 1 | 174 | XP_016860649.1 | |
| AF060798.1 | AAC28337.1 | |||
| AF203910.1 | AAG23728.1 | |||
| AK096954.1 | 1 | 177 | ||
| AK225392.1 | ||||
| AK292694.1 | ||||
| AK298471.1 | ||||
| AK302182.1 | 1 | 174 | ||
| AK310793.1 | 1 | 205 | ||
| AL552406.3 | 1 | 182 | ||
| BC002618.2 | AAH02618.1 | |||
| BC053998.1 | AAH53998.1 | |||
| CB111662.1 | 1 | 214 | ||
| CN268850.1 | 1 | 154 | ||
| DB190729.1 | 1 | 205 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv834543 | Chr.2:219226398 - 219416430 on Build GRCh38 | Loss |
|
| esv3893555 | Chr.2:219172722 - 219273269 on Build GRCh38 | Loss |
|
| nsv508885 | Chr.2:219231225 - 219333602 on Build GRCh38 | Insertion |
|
More Information
Set Membership: |
|

Genomic Map