Assay Details
Target Gene Details
Entrez Gene ID: | 317719 |
Gene Name: | kelch like family member 10 |
Gene Aliases: |
SPGF11 |
Location: |
Chr.17:41835685-41848347 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 5 - Exon 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KLHL10 | NM_152467.3 | NP_689680.2 | ||
| XM_011524706.2 | XP_011523008.1 | |||
| AK057224.1 | BAB71387.1 | |||
| AK301797.1 | ||||
| AY495339.1 | AAS91792.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1057008 | Chr.17:41832887 - 41885423 on Build GRCh38 | Gain |
|
| nsv833447 | Chr.17:41794279 - 41955275 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map