Assay Details
Target Gene Details
Entrez Gene ID: | 112 |
Gene Name: | adenylate cyclase 6 |
Gene Aliases: |
AC6, LCCS8 |
Location: |
Chr.12:48766191-48789096 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 19 - Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ADCY6 | NM_015270.4 | NP_056085.1 | ||
| XM_006719210.3 | XP_006719273.1 | |||
| XM_017018743.1 | XP_016874232.1 | |||
| AB007882.2 | ||||
| AF250226.1 | AAF82478.1 | |||
| AK294446.1 | ||||
| BX537664.1 |
Target Gene Details
Entrez Gene ID: | 100616262 |
Gene Name: | microRNA 4701 |
Gene Aliases: |
- |
Location: |
Chr.12:48771975-48772037 on Build GRCh38 |
Assay Gene Location: | Overlaps - Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MIR4701 | NR_039850.2 |
More Information
Set Membership: |
|

Genomic Map