Assay Details
Target Gene Details
Entrez Gene ID: | 5052 |
Gene Name: | peroxiredoxin 1 |
Gene Aliases: |
MSP23, NKEF-A, NKEFA, PAG, PAGA, PAGB, PRX1, PRXI, TDPX2 |
Location: |
Chr.1:45511035-45522890 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 6 - Exon 7 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDX1 | NM_001202431.1 | NP_001189360.1 | ||
| NM_002574.3 | NP_002565.1 | |||
| NM_181696.2 | NP_859047.1 | |||
| NM_181697.2 | NP_859048.1 | |||
| AB451262.1 | ||||
| AB451388.1 | ||||
| AI912389.1 | ||||
| AK131049.1 | ||||
| AK311898.1 | ||||
| BC007063.1 | AAH07063.1 | |||
| BC021683.1 | AAH21683.1 | |||
| BM740381.1 | ||||
| BT019740.1 | AAV38545.1 | |||
| CR407652.1 | CAG28580.1 | |||
| L19184.1 | AAA50464.1 | |||
| X67951.1 | CAA48137.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3893512 | Chr.1:45476180 - 46068871 on Build GRCh38 | Gain |
|
| nsv829726 | Chr.1:45431909 - 45637514 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map