Assay Details
Target Gene Details
Entrez Gene ID: | 1677 |
Gene Name: | DNA fragmentation factor subunit beta |
Gene Aliases: |
CAD, CPAN, DFF-40, DFF2, DFF40 |
Location: |
Chr.1:3857143-3885429 on Build GRCh38 |
Assay Gene Location: | Overlaps Exon 4 - Intron 4 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1012423 | Chr.1:3477686 - 4747449 on Build GRCh38 | Gain |
|
| esv3585042 | Chr.1:3585575 - 4260083 on Build GRCh38 | Gain |
|
| nsv509590 | Chr.1:3826457 - 3902492 on Build GRCh38 | Insertion |
|
| esv3585052 | Chr.1:3817921 - 3928224 on Build GRCh38 | Gain |
|
| nsv545130 | Chr.1:3826568 - 4283888 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map