Assay Details
Target Gene Details
Entrez Gene ID: | 4968 |
Gene Name: | 8-oxoguanine DNA glycosylase |
Gene Aliases: |
HMMH, HOGG1, MUTM, OGH1 |
Location: |
Chr.3:9749944-9788246 on Build GRCh38 |
Assay Gene Location: | Within Intron 8 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| OGG1 | XM_011533760.1 | XP_011532062.1 | ||
| XM_017006494.1 | XP_016861983.1 | |||
| XM_017006496.1 | XP_016861985.1 | |||
| XM_017006497.1 | XP_016861986.1 | |||
| XM_017006499.1 | XP_016861988.1 |
Target Gene Details
Entrez Gene ID: | 10474 |
Gene Name: | transcriptional adaptor 3 |
Gene Aliases: |
ADA3, NGG1, STAF54, TADA3L, hADA3 |
Location: |
Chr.3:9779964-9793011 on Build GRCh38 |
Assay Gene Location: | Within Exon 9 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TADA3 | NM_001278270.1 | 9 | 1807 | NP_001265199.1 |
| NM_006354.3 | 9 | 2413 | NP_006345.1 | |
| NR_103488.1 | 7 | 1849 | ||
| AA602644.1 | 1 | 127 | ||
| AK000228.1 | 9 | 1817 | ||
| AK024982.1 | 5 | 1769 | ||
| BC013433.1 | 9 | 2076 | AAH13433.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1010768 | Chr.3:9502870 - 9873902 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map