Assay Details
Target Gene Details
Entrez Gene ID: | 539 |
Gene Name: | ATP synthase, H+ transporting, mitochondrial F1 complex, O subunit |
Gene Aliases: |
ATPO, HMC08D05, OSCP |
Location: |
Chr.21:33903453-33915854 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 3 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP5O | NM_001697.2 | NP_001688.1 | ||
| AF088071.1 | ||||
| AK124187.1 | ||||
| AK222608.1 | BAD96328.1 | |||
| AK222962.1 | BAD96682.1 | |||
| AK311796.1 | ||||
| AV734214.1 | ||||
| BC021233.2 | AAH21233.1 | |||
| BC022865.1 | AAH22865.1 | |||
| BT019836.1 | AAV38639.1 | |||
| CD174837.1 | ||||
| CR456822.1 | CAG33103.1 | |||
| X83218.1 | CAA58219.1 | |||
| X83219.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv587399 | Chr.21:33834576 - 33969278 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map