Assay Details
Target Gene Details
Entrez Gene ID: | 51606 |
Gene Name: | ATPase H+ transporting V1 subunit H |
Gene Aliases: |
CGI-11, MSTP042, NBP1, SFD, SFDalpha, SFDbeta, VMA13 |
Location: |
Chr.8:53715543-53843311 on Build GRCh38 |
Assay Gene Location: | Within Exon 15 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| ATP6V1H | NM_015941.3 | 14 | 2118 | NP_057025.2 |
| NM_213619.2 | 13 | 2064 | NP_998784.1 | |
| NM_213620.2 | 14 | 2012 | NP_998785.1 | |
| AF132945.1 | 14 | 2038 | AAD27720.1 | |
| AI741756.1 | 1 | 69 | ||
| AK098305.1 | 14 | 1957 | ||
| BG197001.1 | 1 | 382 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv2759616 | Chr.8:53562029 - 53824913 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map