Assay Details
Target Gene Details
Entrez Gene ID: | 11124 |
Gene Name: | Fas associated factor 1 |
Gene Aliases: |
CGI-03, HFAF1s, UBXD12, UBXN3A, hFAF1 |
Location: |
Chr.1:50441263-50960264 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| FAF1 | NM_007051.2 | 19 | 2480 | NP_008982.1 |
| XM_017000136.1 | 20 | 2197 | XP_016855625.1 | |
| AF094700.1 | 16 | 1553 | AAD51876.1 | |
| AF106798.1 | 19 | 2027 | AAD51886.1 | |
| AF132938.1 | 19 | 2076 | AAD27713.1 | |
| AF136173.1 | 19 | 2076 | AAP97263.1 | |
| AJ271408.1 | 19 | 2352 | CAB67705.1 | |
| AK094843.1 | 14 | 2037 | ||
| AK292840.1 | 20 | 2090 | ||
| AK293659.1 | 12 | 1451 | ||
| AL133631.1 | 16 | 1740 | CAB63755.1 | |
| BC004970.1 | 19 | 2480 | AAH04970.1 | |
| BC067100.1 | 19 | 2304 | AAH67100.1 |
More Information
Set Membership: |
|

Genomic Map