Assay Details
Target Gene Details
Entrez Gene ID: | 4801 |
Gene Name: | nuclear transcription factor Y subunit beta |
Gene Aliases: |
CBF-A, CBF-B, HAP3, NF-YB |
Location: |
Chr.12:104117080-104138262 on Build GRCh38 |
Assay Gene Location: | Within Exon 10 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| NFYB | NM_006166.3 | 8 | 1739 | NP_006157.1 |
| XM_005268908.4 | 9 | 1803 | XP_005268965.1 | |
| XM_005268909.4 | 9 | 1786 | XP_005268966.1 | |
| XM_005268910.4 | 9 | 2458 | XP_005268967.1 | |
| BQ420877.1 | ||||
| CD678671.1 | 1 | 278 |
More Information
Set Membership: |
|

Genomic Map