Assay Details
Target Gene Details
Entrez Gene ID: | 23178 |
Gene Name: | PAS domain containing serine/threonine kinase |
Gene Aliases: |
PASKIN, STK37 |
Location: |
Chr.2:241106099-241150347 on Build GRCh38 |
Assay Gene Location: | Within Exon 23 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PASK | NM_001252119.1 | 18 | 4458 | NP_001239048.1 |
| NM_001252120.1 | 19 | 4590 | NP_001239049.1 | |
| NM_001252122.1 | 18 | 4332 | NP_001239051.1 | |
| NM_015148.3 | 18 | 4437 | NP_055963.2 | |
| XM_005246991.1 | 15 | 3772 | XP_005247048.1 | |
| XM_011510826.1 | 18 | 4413 | XP_011509128.1 | |
| XM_011510828.1 | 19 | 4514 | XP_011509130.1 | |
| XM_011510829.1 | 18 | 4455 | XP_011509131.1 | |
| XM_011510830.1 | 18 | 4456 | XP_011509132.1 | |
| XM_011510831.2 | 16 | 4620 | XP_011509133.1 | |
| XM_011510832.2 | 16 | 4599 | XP_011509134.1 | |
| XM_011510833.1 | 15 | 3793 | XP_011509135.1 | |
| XM_011510834.1 | 11 | 3516 | XP_011509136.1 | |
| XM_011510835.1 | 11 | 3106 | XP_011509137.1 | |
| XM_017003634.1 | 18 | 4681 | XP_016859123.1 | |
| XM_017003635.1 | 19 | 4493 | XP_016859124.1 | |
| XM_017003636.1 | 18 | 4449 | XP_016859125.1 | |
| AF387103.1 | 18 | 4377 | AAK69752.1 | |
| AK296945.1 | 15 | 3942 | ||
| BC063585.1 | 18 | 4380 | AAH63585.1 | |
| U79240.1 | 9 | 1711 | AAB50198.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| esv3575345 | Chr.2:241086207 - 241153068 on Build GRCh38 | Gain |
|
More Information
Additional Information:
For this assay, SNP(s) [rs368917444] are located under a primer or probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Set Membership: |
|

Genomic Map