Assay Details
Target Gene Details
Entrez Gene ID: | 254251 |
Gene Name: | ligand dependent nuclear receptor corepressor like |
Gene Aliases: |
MLR1 |
Location: |
Chr.4:17841199-18023287 on Build GRCh38 |
Assay Gene Location: | Within Exon 13 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| LCORL | NM_153686.8 | 7 | 1471 | NP_710153.2 |
| NR_136669.1 | 5 | 1297 | ||
| XM_011513821.2 | 9 | 6202 | XP_011512123.1 | |
| XM_011513822.2 | 8 | 6175 | XP_011512124.1 | |
| XM_011513823.2 | 8 | 6048 | XP_011512125.1 | |
| XM_011513826.2 | 6 | 1255 | XP_011512128.1 | |
| XM_017007961.1 | 8 | 7484 | XP_016863450.1 | |
| XM_017007962.1 | 8 | 6126 | XP_016863451.1 | |
| XM_017007963.1 | 8 | 6429 | XP_016863452.1 | |
| XM_017007965.1 | 6 | 2530 | XP_016863454.1 | |
| AK096181.1 | 8 | 1332 | ||
| BC037322.1 | 7 | 1354 | AAH37322.3 | |
| BE254250.1 | 1 | 145 |
More Information
Set Membership: |
|

Genomic Map