Assay Details
Target Gene Details
Entrez Gene ID: | 83989 |
Gene Name: | family with sequence similarity 172 member A |
Gene Aliases: |
C5orf21 |
Location: |
Chr.5:93615130-94111699 on Build GRCh38 |
Assay Gene Location: | Within Intron 19 |
Target Gene Details
Entrez Gene ID: | 134187 |
Gene Name: | POU domain class 5, transcription factor 2 |
Gene Aliases: |
SPRM-1 |
Location: |
Chr.5:93740309-93741603 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| POU5F2 | NM_153216.1 | 1 | 674 | NP_694948.1 |
| AK098546.1 | 1 | 674 | BAC05328.1 | |
| BC029532.1 | 1 | 634 | ||
| BX641019.1 | 1 | 194 | CAE46012.1 | |
| Z21965.1 | 1 | 134 | CAA79976.1 |
More Information
Set Membership: |
|

Genomic Map