Assay Details
Target Gene Details
Entrez Gene ID: | 54868 |
Gene Name: | transmembrane protein 104 |
Gene Aliases: |
- |
Location: |
Chr.17:74776483-74839783 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TMEM104 | NM_001321264.1 | 3 | 272 | NP_001308193.1 |
| NM_017728.3 | 3 | 272 | NP_060198.3 | |
| XM_006721963.1 | 3 | 246 | XP_006722026.1 | |
| XM_017024797.1 | 3 | 215 | XP_016880286.1 | |
| XM_017024798.1 | 3 | 246 | XP_016880287.1 | |
| XM_017024799.1 | 3 | 246 | XP_016880288.1 | |
| AK000262.1 | 3 | 255 | BAA91039.1 | |
| AK074029.1 | 3 | 232 | ||
| AK291159.1 | 3 | 267 | ||
| AK296621.1 | ||||
| BC037278.1 | 3 | 224 | AAH37278.1 | |
| DN999695.1 | 3 | 278 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv952367 | Chr.17:74751962 - 74961105 on Build GRCh38 | Deletion |
|
| nsv576025 | Chr.17:74778036 - 74818437 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map