Assay Details
Target Gene Details
Entrez Gene ID: | 440738 |
Gene Name: | microtubule associated protein 1 light chain 3 gamma |
Gene Aliases: |
ATG8J, LC3C |
Location: |
Chr.1:241995194-242001393 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 4 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| MAP1LC3C | NM_001004343.2 | NP_001004343.1 | ||
| XM_005273139.3 | XP_005273196.1 | |||
| AF276659.1 | AAK35152.1 | |||
| BC127722.1 | ||||
| BC132986.1 | ||||
| BC132988.1 | ||||
| CN260933.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv16e55 | Chr.1:241861796 - 242169657 on Build GRCh38 | Gain |
|
| nsv1002309 | Chr.1:241878940 - 242132333 on Build GRCh38 | Gain |
|
| nsv508703 | Chr.1:241983638 - 242088799 on Build GRCh38 | Deletion |
|
| nsv1008886 | Chr.1:241852289 - 242013681 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map