Assay Details
Target Gene Details
Entrez Gene ID: | 5734 |
Gene Name: | prostaglandin E receptor 4 |
Gene Aliases: |
EP4, EP4R |
Location: |
Chr.5:40679930-40740932 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PTGER4 | XM_017009659.1 | XP_016865148.1 |
Target Gene Details
Entrez Gene ID: | 23548 |
Gene Name: | tetratricopeptide repeat domain 33 |
Gene Aliases: |
OSRF |
Location: |
Chr.5:40711576-40756965 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| TTC33 | NM_012382.2 | 5 | 684 | NP_036514.1 |
| XM_011514003.2 | 5 | 1809 | XP_011512305.1 | |
| AF023244.1 | 5 | 630 | AAD09341.1 | |
| AK303119.1 | 4 | 592 | ||
| AK307131.1 | 3 | 445 | ||
| AK312560.1 | 5 | 658 | ||
| BC015701.1 | 5 | 654 | AAH15701.1 | |
| BC036536.1 | 5 | 633 | AAH36536.1 |
More Information
Set Membership: |
|

Genomic Map