Assay Details
Target Gene Details
Entrez Gene ID: | 904 |
Gene Name: | cyclin T1 |
Gene Aliases: |
CCNT, CYCT1, HIVE1 |
Location: |
Chr.12:48688458-48716998 on Build GRCh38 |
Assay Gene Location: | Within Exon 1 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| CCNT1 | NM_001240.3 | 1 | 77 | NP_001231.2 |
| NM_001277842.1 | 1 | 77 | NP_001264771.1 | |
| AK290990.1 | ||||
| HY016640.1 | 1 | 141 |
More Information
Set Membership: |
|

Genomic Map