Assay Details
Target Gene Details
Entrez Gene ID: | 159371 |
Gene Name: | solute carrier family 35 member G1 |
Gene Aliases: |
C10orf60, TMEM20 |
Location: |
Chr.10:93893951-93914265 on Build GRCh38 |
Assay Gene Location: | Within Exon 3 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| SLC35G1 | NM_001134658.1 | 3 | 2035 | NP_001128130.1 |
| NM_153226.2 | 3 | 2032 | NP_694958.1 | |
| XM_011539371.2 | 3 | 2000 | XP_011537673.1 | |
| XM_011539373.2 | XP_011537675.1 | |||
| AK091309.1 | 3 | 2032 | BAC03633.1 | |
| BC041432.1 | 2 | 1764 | AAH41432.1 |
More Information
Set Membership: |
|

Genomic Map