Assay Details
Target Gene Details
Entrez Gene ID: | 25824 |
Gene Name: | peroxiredoxin 5 |
Gene Aliases: |
ACR1, AOEB166, B166, HEL-S-55, PLP, PMP20, PRDX6, PRXV, SBBI10, prx-V |
Location: |
Chr.11:64318088-64321823 on Build GRCh38 |
Assay Gene Location: | Overlaps Intron 4 - Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| PRDX5 | NM_012094.4 | NP_036226.1 | ||
| NM_181651.2 | NP_857634.1 | |||
| NM_181652.2 | NP_857635.1 | |||
| XM_005273882.4 | XP_005273939.2 | |||
| AA187737.1 | ||||
| AF110731.1 | AAF03750.1 | |||
| AF112212.1 | AAF17200.1 | |||
| AF124993.1 | AAF27531.1 | |||
| AF197952.1 | AAF04856.1 | |||
| AF231705.1 | AAF78899.1 | |||
| AF242525.1 | AAF99605.1 | |||
| AJ249483.1 | CAB62210.1 | |||
| BC110983.1 | AAI10984.1 | |||
| BC113723.1 | ||||
| BC113725.1 | ||||
| BC143849.1 | ||||
| BC171733.1 | ||||
| BU598032.1 | ||||
| CR457203.1 | CAG33484.1 | |||
| FJ224298.1 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv951010 | Chr.11:63942829 - 64352128 on Build GRCh38 | Deletion |
|
| dgv1963n54 | Chr.11:64174262 - 64329761 on Build GRCh38 | Loss |
|
| nsv832186 | Chr.11:64204788 - 64402587 on Build GRCh38 | Loss |
|
More Information
Set Membership: |
|

Genomic Map