Assay Details
Target Gene Details
Entrez Gene ID: | 55352 |
Gene Name: | coordinator of PRMT5 and differentiation stimulator |
Gene Aliases: |
C17orf79, COPR5, HSA272196, TTP1 |
Location: |
Chr.17:31851865-31859307 on Build GRCh38 |
Assay Gene Location: | Within Exon 5 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| COPRS | NM_018405.3 | 4 | 604 | NP_060875.2 |
| XM_005258000.1 | 4 | 714 | XP_005258057.1 | |
| AF407672.1 | 4 | 496 | AAK97661.1 | |
| AJ272196.2 | 4 | 511 | CAB77267.2 | |
| AK024772.1 | 4 | 524 | ||
| BC012386.1 | 4 | 534 | AAH12386.1 | |
| BQ671018.1 | 1 | 95 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| nsv1062194 | Chr.17:31755256 - 32041289 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map