Assay Details
Target Gene Details
Entrez Gene ID: | 338599 |
Gene Name: | dual specificity phosphatase and pro isomerase domain containing 1 |
Gene Aliases: |
DUSP27, FMDSP |
Location: |
Chr.10:75026427-75073827 on Build GRCh38 |
Assay Gene Location: | Within Intron 6 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| DUPD1 | XM_011539747.2 | XP_011538049.1 |
Target Gene Details
Entrez Gene ID: | 23522 |
Gene Name: | lysine acetyltransferase 6B |
Gene Aliases: |
GTPTS, MORF, MOZ2, MYST4, ZC2HC6B, qkf, querkopf |
Location: |
Chr.10:74824927-75032623 on Build GRCh38 |
Assay Gene Location: | Within Exon 20 |
| Gene Symbol | Transcript Accession | Exon Location | Assay Transcript Location | Protein ID |
|---|---|---|---|---|
| KAT6B | NM_001256468.1 | 18 | 6210 | NP_001243397.1 |
| NM_001256469.1 | 18 | 5883 | NP_001243398.1 | |
| NM_012330.3 | 18 | 6759 | NP_036462.2 | |
| XM_005269664.2 | 18 | 6820 | XP_005269721.1 | |
| XM_017016000.1 | 18 | 6668 | XP_016871489.1 | |
| XM_017016001.1 | 18 | 6532 | XP_016871490.1 | |
| XM_017016002.1 | 18 | 6771 | XP_016871491.1 | |
| XM_017016003.1 | 17 | 6442 | XP_016871492.1 | |
| XM_017016004.1 | 17 | 6506 | XP_016871493.1 | |
| XM_017016005.1 | 18 | 6119 | XP_016871494.1 | |
| XM_017016006.1 | 18 | 5792 | XP_016871495.1 | |
| XM_017016007.1 | 18 | 5655 | XP_016871496.1 | |
| XM_017016008.1 | 17 | 5566 | XP_016871497.1 | |
| XM_017016009.1 | 17 | 5721 | XP_016871498.1 | |
| AB002381.2 | 16 | 5182 | BAA20837.2 | |
| AF113514.1 | 17 | 5497 | AAF00095.1 | |
| AF119230.1 | 17 | 5824 | AAF00099.1 | |
| AF119231.1 | 17 | 6373 | AAF00100.1 | |
| AF217500.1 | 18 | 6543 | AAL56647.1 | |
| BC021128.1 | 2 | 2501 | AAH21128.1 | |
| BC150270.1 | 16 | 5209 | ||
| BC150618.1 | 18 | 6519 |
Target Copy Number Variation Details
DGV Version: |
Release date: 2016-05-15, GRCh GRCh38 |
| Target Variation |
Location | CNV Subtype |
Genes |
|---|---|---|---|
| dgv926n100 | Chr.10:74994872 - 75081593 on Build GRCh38 | Gain |
|
| dgv927n100 | Chr.10:75024700 - 75329039 on Build GRCh38 | Gain |
|
More Information
Set Membership: |
|

Genomic Map